Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Focal Adhesion Kinase Mouse anti-Human, Mouse, Rat, Chicken, Clone: BLAb2H7, MilliporeSigma™

Catalog No. 051139MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-113-9MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-113-9MI Supplier MilliporeSigma Supplier No. 051139
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Focal Adhesion Kinase Antibody, clone BLAb2H7 is validated for use in western blot, immunoprecipitation for the detection of Focal Adhesion Kinase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Focal Adhesion Kinase
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone BLAb2H7
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1 in buffer containing 0.05% sodium azide.
Gene Accession No. NP_990766.1
Gene Symbols EC 2.7.10.2, FADK, FAK , FAK1 , pp125FAK
Host Species Mouse
Immunogen Recombinant fragment derived from the amino terminal region of chicken FAK, amino acid residues 103 to 553.
Quantity 100 μg
Primary or Secondary Primary
Target Species Chicken, Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.