Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Fodrin Clone: Fod009,

Catalog No. 34405050UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
34-405-050UG 50 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-405-050UG Supplier MilliporeSigma™ Supplier No. 34405050UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Fodrin Clone: Fod009 in Bovine, Canine, Human, Mouse, Porcine, Rabbit, Rat samples, and it is validated for ELISA, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immuno Blotting

Fodrin is a spectrin-like cytoskeletal protein found in non-erythroid cells and is comprised of two subunits, α and β (∽240kDa and ∽235kDa, respectively). It mediates the association of actin filaments with the plasma membrane and may thus be involved in maintenance of cell shape and stabilization of membrane structure. Fodrin's susceptibility to tyrosine phosphorylation and its interaction with the lymphocyte transmembrane protein tyrosine phosphatase CD45 suggest that it plays an important role in signal transduction during lymphocyte activation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fodrin
Applications ELISA, Flow Cytometry, Immunoblot, Immunoblot, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone Fod009
Conjugate Unconjugated
Formulation In 150mM NaCl, 10mM sodium phosphate, pH 7.5.
Host Species Mouse
Immunogen purified bovine brain fodrin
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽240kDa intracellular and plasma membrane associated β subunit of fodrin.
Target Species Bovine, Canine, Human, Mouse, Pig, Rabbit, Rat
Content And Storage ≤ -70°C
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.