Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Follicular Dendritic Cell Rat anti-Mouse, Unlabeled, Clone: FDC-M1, BD

Catalog No. BDB551320 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB551320 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB551320 Supplier BD Biosciences Supplier No. 551320
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Follicular Dendritic Cells (FDC) form a structural network within the lymphoid follicles and are involved in antigen presentation to B cells for generating an immune response and for the maintenance of immunological memeory. FDCs are believed to provide signals to B cells to induce their proliferation. In LCMV mediated immunosuppression there is evidence of T-cell dependent destruction of macrophages and follicular dendritic cells. The FDC-M1 antibody recognizes follicular dendritic cells and their dendritic processes.

Host Species: Rat
Clone: FDC-M1
Isotype: IgG2c, κ
Species Reactivity: Mouse

Immunohistochemistry, Immunohistochemistry (Frozen)

TRUSTED_SUSTAINABILITY

Specifications

Antigen Follicular Dendritic Cell
Applications Immunohistochemistry (Frozen)
Classification Monoclonal
Clone FDC-M1
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, goat serum, and ≤0.09% sodium azide.
Host Species Rat
Purification Method Affinity Purified
Quantity 1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG2c κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.