Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Folliculin (FLCN), Mouse anti-Human, Clone: 8D9.2, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABC288
Description
This Anti-Folliculin (FLCN), Antibody clone 8D9.2 is validated for use in Western Blotting and Immunocytochemistry for the detection of Folliculin (FLCN).
Specifications
Folliculin (FLCN) | |
Monoclonal | |
Unconjugated | |
NP_653207 | |
Mouse | |
Purified by ion-exchange chromatography | |
Primary | |
2-8°C for one year | |
IgM κ |
Immunocytochemistry, Western Blot | |
8D9.2 | |
Purified mouse monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide. | |
FLCN, BHD | |
GST-tagged recombinant protein corresponding to human Folliculin (FLCN). | |
100 μg | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction