Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fra2 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA535174
Description
This antibody is predicted to react with mouse and rat based on sequence homology.
The Fos gene family consists of 4 members: FOS, FOSB, FOSL1, and FOSL2. These genes encode leucine zipper proteins that can dimerize with proteins of the JUN family, thereby forming the transcription factor complex AP-1. As such, the FOS proteins have been implicated as regulators of cell proliferation, differentiation, and transformation.
Specifications
| Fra2 | |
| Polyclonal | |
| PBS with 0.09% sodium azide | |
| P15408 | |
| FOSL2 | |
| KLH conjugated synthetic peptide between 220-248 amino acids from the C-terminal region of human FOSL2 | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| Unconjugated | |
| FOSL2 | |
| FOSL2; FRA2; Fos-related antigen 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2355 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction