Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FOXC1 Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABD71
Description
Anti-FOXC1 Antibody is a Rabbit Polyclonal Antibody for detection of FOXC1 also known as Forkhead box protein C1, Forkhead-related protein FKHL7, FREAC-3 and has been validated in western blot, immunocytochemistry.
Specifications
FOXC1 | |
Polyclonal | |
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
Rabbit | |
Affinity Purified | |
Primary | |
Human, Mouse |
Immunocytochemistry, Western Blot | |
Unconjugated | |
FOXC1; FKHL7; FREAC3 | |
KLH-conjugated linear peptide corresponding to human FOXC1. | |
100 μg | |
NP_001444 | |
2-8°C for 12 months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction