Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXC2, Mouse, Clone: 3E6, Abnova™

Catalog No. 89022147 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-022-147 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-022-147 Supplier Abnova Corporation Supplier No. H00002303M08
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant FOXC2.

This gene belongs to the forkhead family of transcription factors which is characterized by a distinct DNA-binding forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in the development of mesenchymal tissues. [provided by RefSeq

Sequence: FENGSFLRRRRRFKKKDVSKEKEERAHLKEPPPAASKGAPATPHLADAPKEAEKKVVIKSEAASPALPVITKVETLSPESALQGSPRSAASTPAGSPDGSL

Specifications

Antigen FOXC2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3E6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant FOXC2.
Formulation PBS with no preservative; pH 7.4
Gene FOXC2
Gene Accession No. NM_005251
Gene Alias FKHL14/LD/MFH-1/MFH1
Gene Symbols FOXC2
Host Species Mouse
Immunogen FOXC2 (NP_005242, 156 a.a. ∼ 256 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2303
Target Species Human, Rat
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.