Learn More
FOXO3, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human FOXO3 peptide using ARM Technology.
Supplier: Abnova Corporation H00002309K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
This gene belongs to the forkhead family of transcription factors which are characterized by a distinct forkhead domain. This gene likely functions as a trigger for apoptosis through expression of genes necessary for cell death. Translocation of this gene with the MLL gene is associated with secondary acute leukemia. Alternatively spliced transcript variants encoding the same protein have been observed. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
| FOXO3 | |
| Monoclonal | |
| Unconjugated | |
| In 1X PBS, pH 7.4 | |
| FOXO3 | |
| FOXO3 | |
| A synthetic peptide of human FOXO3 | |
| RUO | |
| 2309 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| ELISA, Western Blot | |
| 1mg | |
| forkhead box O3 | |
| FOXO3 | |
| AF6q21, DKFZp781A0677, FKHRL1, FKHRL1P2, FOXO2, FOXO3A, MGC12739, MGC31925 | |
| Rabbit | |
| 1 mg x up to 3 Abs | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.