Learn More
Description
This gene belongs to the forkhead family of transcription factors which are characterized by a distinct forkhead domain. This gene likely functions as a trigger for apoptosis through expression of genes necessary for cell death. Translocation of this gene with the MLL gene is associated with secondary acute leukemia. Alternatively spliced transcript variants encoding the same protein have been observed. [provided by RefSeq
Specifications
Specifications
| Antigen | FOXO3A |
| Applications | ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot |
| Classification | Monoclonal |
| Clone | 2C4 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against a partial recombinant FOXO3A. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | FOXO3 |
| Gene Accession No. | BC021224 |
| Gene Alias | AF6q21/DKFZp781A0677/FKHRL1/FKHRL1P2/FOXO2/FOXO3A/MGC12739/MGC31925 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.