Learn More
FOXP1 Rabbit anti-Human, Mouse, Rat, Bovine, Rhesus Macaque, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB2277
Description
Anti-FOXP1 Antibody detects level of FOXP1 and has been published and validated for use in western blot, immunocytochemistry, immunohistochemistry.
Specifications
| FoxP1 | |
| Polyclonal | |
| Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide. | |
| Rabbit | |
| Affinity Purified | |
| Primary | |
| Bovine, Human, Mouse, Rat, Rhesus Monkey |
| Immunocytochemistry, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| FOXP1; QRF1; 12CC4; HSPC215; hFKH1B | |
| KLH-conjugated linear peptide corresponding to the C-terminus of human FOXP1. | |
| 100 μg | |
| NP_116071 | |
| 2-8°C for 12 months |