Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FoxP2 Rabbit anti-Mouse, Rabbit, Human, Polyclonal, MilliporeSigma™

Catalog No. ABE73MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE73MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE73MI Supplier MilliporeSigma Supplier No. ABE73
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-FoxP2 Antibody is a Rabbit Polyclonal Antibody for detection of FoxP2 also known as forkhead box P2, trinucleotide repeat containing 10 and has been validated in western blotting and immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FoxP2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Unpurified rabbit polyclonal serum with 0.05% sodium azide.
Gene Symbols CAGH44; TNRC10; SPCH1
Host Species Rabbit
Immunogen GST-tagged recombinant protein corresponding to mouse FoxP2.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_444472
Target Species Human, Mouse, Rabbit
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.