Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Foxp3 Rat anti-Mouse, Alexa Fluor 488, Clone: MF23, BD

Catalog No. BDB560403 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560403 0.1 mg
BDB560407 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB560403 Supplier BD Biosciences Supplier No. 560403
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Foxp3 is a 50-55kDa protein also known as Forkhead box P3, JM2, or IPEX. It is a member of the forkhead or winged helix family of transcription factors and is specifically expressed by T regulatory (Treg) cells. Foxp3 has been reported to be a key regulatory protein for Treg cell development and function. Ectopic expression of Foxp3 in conventional T cells is sufficient to induce suppressive activity, repress the production of cytokines such as IL2 and IFN-γ, and upregulate Treg cell-associated molecules such as CD25, CTLA4 and GITR. It has been found that the mutation of Foxp3 is responsible for “scurfy” mice. When overexpressed, Foxp3 leads to poor T cell proliferation and activation.
Monoclonal antibody MF23 reacts with mouse Foxp3. Immunoblotting with MF23 antibody has confirmed it recognizes an epitope between 1-87 amino acids in the N-terminal domain.

Host Species: Rat
Clone: MF23
Isotype: IgG2b
Species Reactivity: Mouse
Immunogen: Mouse Foxp3 Recombinant Protein

Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen Foxp3
Applications Flow Cytometry
Classification Monoclonal
Clone MF23
Concentration 0.2mg/mL
Conjugate Alexa Fluor 488
Description Forkhead box p3; Forkhead box protein p3; JM2; Scurfin; Scurfy; Sf
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse Foxp3 Recombinant Protein
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.