Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Foxp3 Mouse anti-Mouse, APC, Clone: 3G3, MilliporeSigma™

Catalog No. MABF462MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF462MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF462MI Supplier MilliporeSigma Supplier No. MABF462
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Foxp3 Antibody,clone 3G3, APC conjugated, is validated for use in Flow Cytometry for the detection of Foxp3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Foxp3
Applications Flow Cytometry
Classification Monoclonal
Clone 3G3
Concentration 0.2mg/mL
Conjugate APC
Formulation Purified Mouse Monoclonal Antibody in buffer containing 10mM NaH2PO4, 150mM NaCl, 0.1% gelatin, pH 7.2, and up to 0.1% sodium azide.
Gene Symbols PFoxp3
Host Species Mouse
Immunogen Proprietary immunogen
Purification Method Size Exclusion Chromatography
Quantity 100 μg
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2-8°C, protect from prolonged exposure to light. Do not Freeze.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.