Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-Foxp3; Clone: R16-715; BD
Supplier: BD Biosciences 563101
Description
Host Species: Rat
Clone R16-715
Isotype IgG2a, κ
Species Reactivity Ms
Immunogen Recombinant Protein
Flow Cytometry, Intracellular Staining

Specifications
Foxp3 | |
R16-715 | |
PE | |
Recombinant Protein | |
100 μg | |
Primary | |
IgG2a |
Monoclonal | |
0.2mg/mL | |
Rat | |
Affinity Purified | |
RUO | |
Mouse |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction