Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FoxP3 Mouse anti-Human, PE, Clone: 236A/E7, BD

Catalog No. BDB560852 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560852 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560852 Supplier BD Biosciences Supplier No. 560852
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 236A/E7 antibody reacts with the human FoxP3 (Forkhead box protein P3) transcription factor, a member of the forkhead or winged helix
family of transcription factors. The expression of FoxP3, also known as Scurfin, IPEX and JM2, has been found to be associated with CD4+
CD25+ regulatory T cells and represents a specific marker for these cells. Flow cytometric analysis has shown that FoxP3 is expressed by the
majority of CD4+ CD25+ (high) T cells in peripheral blood while less than half of CD4+ CD25+ (intermediate) cell population are FoxP3
positive. Approximately 5-10% of peripheral CD4+ cells are CD4+ CD25+ T regulatory cells. T regulatory cells are thought to play a critical
role in the control of T cell mediated autoimmunity by suppressing the proliferation and cytokine production of other T cells. To support this
hypothesis, it has been found that Foxp3 is mutated in scurfy (sf) mice. Cumulative evidence suggests that the 236A/E7 antibody recognizes
epitopes from both isoforms of Exon 2 alternatively-spliced variants and is located in the region between Exon 2 and the Zn/LZ domains (aa
105-235). The antibody was conjugated with R-PE under optimum conditions, and unconjugated antibody and free PE were removed.

Host Species: Mouse
Clone: 236A/E7
IsoType: Isotype: Mouse (BALB/c) IgG1κ
Immunogen: Human full-length FoxP3 Recombinant Protein
Cross Reactivity: Human

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen FoxP3
Applications Flow Cytometry
Classification Monoclonal
Clone 236A/E7
Conjugate PE
Description Forkhead box protein P3; Scurfin; AIID; IPEX; JM2; DIETER; PIDX; XPID
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human full-length FoxP3 Recombinant Protein
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.