Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FoxP3 Mouse anti-Human, V450, Clone: 259D/C7, BD

Catalog No. BDB560459 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
120 Tests
30 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560459 120 Tests
BDB560460 30 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB560459 Supplier BD Biosciences Supplier No. 560459
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 259D/C7 antibody reacts with the human FoxP3 transcription factor, a member of the forkhead or winged helix family of transcription factors. The expression of FoxP3, also known as Scurfin, IPEX and JM2, has been found to be associated with CD4+ regulatory T cells and represents a specific marker for these cells. Flow-cytometric analysis has shown that FoxP3 is expressed by the majority of CD4+CD25+high T cells in peripheral blood while less than half of CD4+CD25int cell population are FoxP3 positive. Approximately 5-10% of peripheral CD4+ cells are CD4+CD25+ T regulatory cells. T regulatory cells are thought to play a critical role in the control of T cell mediated
autoimmunity by suppressing the proliferation and cytokine production of other T cells. To support this hypothesis, it has been found that FOXP3 is mutated in scurfy (sf) mice. The 259D/C7 antibody reacts with all currently identified isoforms of human FoxP3 and is cross-reactive with Cynomolgus, Rhesus and Baboon. The antibody is conjugated to BD Horizon™ V450, which has been developed for use in multicolor flow cytometry experiments and is available exclusively from BD Biosciences. It is excited by the Violet laser Ex max of 406nm and has an Em Max at 450nm. Conjugates with BD Horizon™ V450 can be used in place of Pacific Blue™ conjugates.

Host Species: Mouse
Clone: 259D/C7
Isotype: IgG1
Species Reactivity: Human
Immunogen: FoxP3 recombinant protein

Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen FoxP3
Applications Flow Cytometry
Classification Monoclonal
Clone 259D/C7
Conjugate Violet Fluor 450 (V450)
Description Scurfin, IPEX, JM2
Formulation Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Host Species Mouse
Immunogen FoxP3 recombinant protein
Purification Method Affinity Purified
Quantity 120 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Baboon, Cynomolgus, Human, Rhesus Monkey
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.