Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FoxP4 Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. ABE74MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE74MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE74MI Supplier MilliporeSigma Supplier No. ABE74
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Use Anti-FoxP4 Antibody (Rabbit Polyclonal Antibody) validated in immunocytochemistry, western blot, immunohistochemistry to detect FoxP4 also known as forkhead box P4, winged-helix repressor FOXP4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FoxP4
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Unpurified rabbit polyclonal serum with 0.05% sodium azide.
Gene Symbols Foxp4; FKHLA; hFKHLA
Host Species Rabbit
Immunogen GST-tagged recombinant protein corresponding to mouse FoxP4.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_001104294
Target Species Human, Mouse
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.