Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fractin Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB3150MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB3150MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB3150MI Supplier MilliporeSigma Supplier No. AB3150
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect Fractin using this Anti-Fractin Antibody, C-terminus validated for use in western blot, immunocytochemistry, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fractin
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Liquid whole rabbit serum containing no preservatives.
Gene Symbols ACTB; PS1TP5BP1; beta-actin; Beta-actin
Host Species Rabbit
Immunogen K-YELPD peptide representing the last five amino acids of the C-terminus of the 32 kDa Fractin fragment of beta-Actin. This sequence is absolutely conserved in Fractin fragments from human, mouse, rat, feline, porcine, ovine, bovine, canine, rabbit and gu
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_001101.2
Target Species Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.