Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fragile X Mental Retardation Protein Mouse anti-Human, Mouse, Rat, Clone: 1C3, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB2160
Description
Anti-Fragile X Mental Retardation Protein Antibody, clone 1C3 detects level of Fragile X Mental Retardation Protein and has been published and validated for use in ELISA, immunocytochemistry, immunohistochemistry, immunohistochemistry (paraffin) and western blotting.
Specifications
Fragile X Mental Retardation Protein | |
Monoclonal | |
Unconjugated | |
NM_002024.3 | |
Mouse | |
Unpurified | |
Primary | |
−20°C for up to 12 months. | |
IgG1 κ |
ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot | |
1C3 | |
Ascites mouse monoclonal IgG1k fluid containing no preservatives | |
FMR1, FRAXA, FMRP, MGC87458 | |
Fusion protein with a full length FMRP (human). | |
100 μL | |
Human, Mouse, Rat | |
Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction