Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frataxin Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB15080MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB15080MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB15080MI Supplier MilliporeSigma Supplier No. AB15080
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-Frataxin Antibody is validated for use in western blotting for the detection of Frataxin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Frataxin
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit polyclonal serum in buffer containing 0.05% NaN3.
Host Species Rabbit
Immunogen GST-tagged Frataxin corresponding to mature chain.
Purification Method GST-depleted sera.
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_000135
Target Species Human, Mouse, Rat
Content And Storage −20°C for up to 12 months
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.