Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FRG2B Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA524547
Description

Specifications
FRG2B | |
Polyclonal | |
FRG2B | |
FSHD region gene 2 protein family member B, HSA10-FRG2, protein FRG2-like-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
441581 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
Unconjugated | |
Q96QU4 | |
FRG2B | |
KLH conjugated synthetic peptide between 15-43 amino acids from the N-terminal region of human FRG2B | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction