Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FRMPD2, Rabbit, Monoclonal Antibody, Abnova™

Catalog No. 89204530 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg x up to 3 Abs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-204-530 1 mg x up to 3 Abs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-204-530 Supplier Abnova Corporation Supplier No. H00143162K

Includes

Up to three rabbit IgG clones of 1mg each will be delivered to customer.

Rabbit monoclonal antibody raised against a human FRMPD2 peptide using ARM Technology.

This gene encodes a FERM and PDZ domain-containing protein and is located in a region of chromosome 10q which contains a segmental duplication resulting in three nearly identical regions. This copy of the gene is full-length and is in the telomeric duplicated region. The exact function of this gene is unknown; however, FERM domains often play a role in signal transduction pathways. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq

Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)
Expression: Overexpression vector and transfection into 293H cell line

Order Info

  1. Customer may provide cell or tissue lysate for antibody screening.
  2. Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.

Specifications

Antigen FRMPD2
Applications ELISA, Western Blot
Classification Monoclonal
Concentration 1mg
Conjugate Unconjugated
Description FERM and PDZ domain containing 2
Formulation In 1X PBS, pH 7.4
Gene FRMPD2
Gene Accession No. FRMPD2
Gene Alias MGC35285, MGC87776, MGC87777, MGC90186, PDZD5C, PDZK5C
Gene Symbols FRMPD2
Host Species Rabbit
Immunogen A synthetic peptide of human FRMPD2
Quantity 1 mg x up to 3 Abs
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 143162
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.