Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FRS2/SNT-1 Mouse anti-Human, Rat, MilliporeSigma™

Catalog No. 05502MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-502-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-502-MI Supplier MilliporeSigma Supplier No. 05502
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect FRS2/SNT-1 using this Anti-FRS2/SNT-1 Antibody validated for use in immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FRS2/SNT-1
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation 0.07M Tris-glycine, pH 7.4, 0.105M NaCl, 0.035% sodium azide containing 30% glycerol
Gene Accession No. NM_006654
Gene Symbols FRS2, SNT1, FRS2A, SNT-1, SNT, FRS2alpha
Host Species Mouse
Immunogen Peptide corresponding to residues 283-297 of human FRS2/SNT-1
Purification Method Protein G chromatography
Quantity 200 μg
Primary or Secondary Primary
Target Species Human, Rat
Content And Storage −20°C for 24 months.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.