Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FRS3/FRS2Β Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. 071825MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-182-5MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-182-5MI Supplier MilliporeSigma Supplier No. 071825

Rabbit Polyclonal Antibody

Anti-FRS3/FRS2β Antibody detects level of FRS3/FRS2β and has been published and validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FRS3/FRS2β
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit serum in PBS containing 0.05% sodium azide.
Gene Symbols FRS2-beta ; FRS2B; FRS2beta; MGC17167 ; OTTHUMP00000016383; OTTHUMP00000039786 ; SNT-2; SNT2
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to amino acids 361-375 of FRS3/ FRS2β.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_659188
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.