Learn More
Description
Specifications
Specifications
| Antigen | FSP1/S100A4 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Purified rabbit polyclonal containing PBS with 0.05% sodium azide. |
| Gene Symbols | S100A4; CAPL; FSP1; 18A2; PEL98; P9KA; 42A; MTS1 |
| Host Species | Rabbit |
| Immunogen | Histidine-tagged recombinant protein corresponding to mouse FSP1/S100-A4. |
| Purification Method | Protein G purfied |
| Quantity | 100 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.