Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FTL, Mouse, Clone: 4G9, Abnova™

Catalog No. 89022228 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-022-228 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-022-228 Supplier Abnova Corporation Supplier No. H00002512M10
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length native FTL.

This gene encodes the light subunit of the ferritin protein. Ferritin is the major intracellular iron storage protein in prokaryotes and eukaryotes. It is composed of 24 subunits of the heavy and light ferritin chains. Variation in ferritin subunit composition may affect the rates of iron uptake and release in different tissues. A major function of ferritin is the storage of iron in a soluble and nontoxic state. Defects in this light chain ferritin gene are associated with several neurodegenerative diseases and hyperferritinemia-cataract syndrome. This gene has multiple pseudogenes. [provided by RefSeq

Specifications

Antigen FTL
Applications ELISA
Classification Monoclonal
Clone 4G9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length native FTL.
Dilution The optimal working dilution should be determined by the end user.
Formulation PBS with no preservative; pH 7.4
Gene FTL
Gene Accession No. P02792
Gene Alias MGC71996
Gene Symbols FTL
Host Species Mouse
Immunogen Native purified human FTL.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2512
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.