Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FtsJ homolog 1 (E. coli), Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89007333 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-007-333 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-007-333 Supplier Abnova Corporation Supplier No. H00024140B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human FTSJ1 protein.

The protein encoded by this gene is a member of the S-adenosylmethionine-binding protein family. It is a nucleolar protein and may be involved in the processing and modification of rRNA. Three alternatively spliced transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq

Sequence: MGRTSKDKRDVYYRLAKENGWRARSAFKLLQLDKEFQLFQGVTRAVDLCAAPGSWSQVLSQKIGGQGSGHVVAVDLQAMAPLPGVVQIQGDITQLSTAKEIIQHFKGCPADLVVCDGAPDVTDLHDVDEYMQAQLLLAALNIATHVLKPGGCFVAKIFRGRDVTLLYSQLQVFFSSVLCAKPRSSRNSSIEAFAVCQGYDPPEGFIPDLSKPLLDHSYDPDFNQLDGPTRIIVPFVTCGDLSSYDSDRSYPLDLEGGSEYKYTPPTQPPISPPYQEACTLKRKGQLAKEIRPQDCPISRVDTFPQPLAAPQCHTLLAPEMEDNEMSCSP

Specifications

Antigen FtsJ homolog 1 (E. coli)
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human FTSJ1 protein.
Formulation No additive
Gene FTSJ1
Gene Accession No. BC023584
Gene Alias CDLIV/JM23/MRX44/MRX9/SPB1/TRM7
Gene Symbols FTSJ1
Host Species Mouse
Immunogen FTSJ1 (AAH23584, 1 a.a. ∼ 329 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 24140
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.