Learn More
FUT3, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human FUT3 peptide using ARM Technology.
Supplier: Abnova Corporation H00002525K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
The Lewis histo-blood group system comprises a set of fucosylated glycosphingolipids that are synthesized by exocrine epithelial cells and circulate in body fluids. The glycosphingolipids function in embryogenesis, tissue differentiation, tumor metastasis, inflammation, and bacterial adhesion. They are secondarily absorbed to red blood cells giving rise to their Lewis phenotype. This gene is a member of the fucosyltransferase family, which catalyzes the addition of fucose to precursor polysaccharides in the last step of Lewis antigen biosynthesis. It encodes an enzyme with alpha(1,3)-fucosyltransferase and alpha(1,4)-fucosyltransferase activities. Mutations in this gene are responsible for the majority of Lewis antigen-negative phenotypes. Multiple alternatively spliced variants, encoding the same protein, have been found for this gene. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
FUT3 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
FUT3 | |
FUT3 | |
A synthetic peptide of human FUT3 | |
RUO | |
2525 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
fucosyltransferase 3 (galactoside 3(4)-L-fucosyltransferase, Lewis blood group) | |
FUT3 | |
CD174, FT3B, FucT-III, LE, Les, MGC131739 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.