Learn More
fucosyltransferase 7 (alpha (1,3) fucosyltransferase), Mouse, Clone: 1C12, Abnova™
Mouse monoclonal antibody raised against a partial recombinant FUT7.
Supplier: Abnova Corporation H00002529M03
Description
The protein encoded by this gene is a Golgi stack membrane protein that is involved in the creation of sialyl-Lewis X antigens. The encoded protein can direct the synthesis of the E-selectin-binding sialyl-Lewis X moiety. [provided by RefSeq
Sequence: YEAFVPADAFVHVDDFGSARELAAFLTGMNESRYQRFFAWRDRLRVRLFTDWRERFCAICDRYPHLPRSQVYEDLEGWFQASpecifications
fucosyltransferase 7 (alpha (1,3) fucosyltransferase) | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_004479 | |
FUT7 | |
FUT7 (NP_004470, 262 a.a. ∼ 342 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Yes | |
2529 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG2a κ |
ELISA, Western Blot | |
1C12 | |
Mouse monoclonal antibody raised against a partial recombinant FUT7. | |
FUT7 | |
FucT-VII | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.