Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FUT9, Rabbit, Monoclonal Antibody, Abnova™

Catalog No. 89201562 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg x up to 3 Abs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-201-562 1 mg x up to 3 Abs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-201-562 Supplier Abnova Corporation Supplier No. H00010690K

Includes

Up to three rabbit IgG clones of 1mg each will be delivered to customer.

Rabbit monoclonal antibody raised against a human FUT9 peptide using ARM Technology.

FUT9 is one of several alpha-3-fucosyltransferases that can catalyze the last step in the biosynthesis of Lewis antigen, the addition of a fucose to precursor polysaccharides. FUT9 synthesizes the LeX oligosaccharide (CD15), which is expressed in organ buds progressing in mesenchyma during human embryogenesis.[supplied by OMIM

Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)
Expression: Overexpression vector and transfection into 293H cell line

Order Info

  1. Customer may provide cell or tissue lysate for antibody screening.
  2. Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.

Specifications

Antigen FUT9
Applications ELISA, Western Blot
Classification Monoclonal
Concentration 1mg
Conjugate Unconjugated
Description fucosyltransferase 9 (alpha (1,3) fucosyltransferase)
Formulation In 1X PBS, pH 7.4
Gene FUT9
Gene Accession No. FUT9
Gene Alias Fuc-TIX
Gene Symbols FUT9
Host Species Rabbit
Immunogen A synthetic peptide of human FUT9
Quantity 1 mg x up to 3 Abs
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10690
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.