Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FXR-B Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB10305
Description
Detect FXR-B using this Anti-FXR-B Antibody validated for use in western blotting.
Specifications
FXR-B | |
Polyclonal | |
Raw rabbit serum with 0.1% sodium azide. | |
Rabbit | |
100 μL | |
NM_005123.2 | |
2-8°C for 12 months | |
IgG2b |
Western Blot | |
Unconjugated | |
NR1H4; HRR1; MGC163445; FXR; HRR-1; RIP14; BAR; FXRA | |
Recombinant human hFXR-B | |
Primary | |
Human | |
Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction