Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FXR1 Mouse anti-Human, Mouse, Clone: 6BG10, MilliporeSigma™

Catalog No. 051529MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-152-9MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-152-9MI Supplier MilliporeSigma Supplier No. 051529
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Use Anti-FXR1 Antibody, clone 6BG10 (Mouse Monoclonal Antibody) validated in western blot, immunocytochemistry, immunoprecipitation to detect FXR1 also known as fragile X mental retardation autosomal homolog 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FXR1
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 6BG10
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
Gene Accession No. NP_001013456.1
Gene Symbols FXR1, hFXR1
Host Species Mouse
Immunogen Recombinant protein corresponding to human FXR1.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.