Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FXR2 Mouse anti-Human, Rat, Mouse, Clone: 2C8.2, MilliporeSigma™

Catalog No. 051214MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-121-4MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-121-4MI Supplier MilliporeSigma Supplier No. 051214

Mouse Monoclonal Antibody

Anti-FXR2 Antibody, clone 2C8.2 is a Mouse Monoclonal Antibody for detection of FXR2 also known as Fragile X mental retardation syndrome-related protein 2 and has been validated in western blot, immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FXR2
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 2C8.2
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_004851
Gene Symbols FXR2, FMR1L2
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to human FXR2.
Purification Method Protein G
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.