Learn More
frizzled homolog 3 (Drosophila), Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant FZD3.
Supplier: Abnova Corporation H00007976A01
Description
This gene is a member of the frizzled gene family. Members of this family encode seven-transmembrane domain proteins that are receptors for the Wingless type MMTV integration site family of signaling proteins. Most frizzled receptors are coupled to the beta-catenin canonical signaling pathway. The function of this protein is unknown, although it may play a role in mammalian hair follicle development. [provided by RefSeq
Sequence: QQTAALAMEPFHPMVNLDCSRDFRPFLCALYAPICMEYGRVTLPCRRLCQRAYSECSKLMEMFGVPWPEDMECSRFPDCDEPYPRLVDLNLAGEPTEGAPVAVSpecifications
frizzled homolog 3 (Drosophila) | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant FZD3. | |
FZD3 | |
Fz-3/hFz3 | |
Mouse | |
50 μL | |
Membrane Receptors | |
Primary | |
Human | |
Serum |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_017412 | |
FZD3 | |
FZD3 (NP_059108, 55 a.a. ∼ 157 a.a) partial recombinant protein with GST tag. | |
RUO | |
Yes | |
7976 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.