Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G-CSF Rat anti-Human, Unlabeled, Clone: BVD13-3A5, BD

Catalog No. BDB551342 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB551342 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB551342 Supplier BD Biosciences Supplier No. 551342
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The BVD13-3A5 antibody reacts with human granulocyte-colony stimulating factor (G-CSF). The immunogen used to generate the hybridoma was E. coli-expressed human G-CSF. This is a neutralizing antibody.

Host Species: Rat
Clone: BVD13-3A5
Isotype: IgG1
Species Reactivity: Human
Immunogen: E. coli-expressed human G-CSF

ELISA, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen G-CSF
Applications ELISA
Classification Monoclonal
Clone BVD13-3A5
Concentration 1.0mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen E. coli-expressed human G-CSF
Purification Method Affinity Purified
Quantity 1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.