Learn More
Description
GTP-binding proteins (G-proteins) play a critical role in signal transduction by coupling receptors to effectors. The intracellular consequences of G-protein activation include second messenger generation, protein phosphorylation, ion channel activation, gene induction, cell growth, and differentiation. Go belongs to the pertussis toxin sensitive G-proteins. It consists of two subtypes of α-subunits (αo1 and αo2) derived from alternative splicing of a single gene. αo1 differs from αo2 within the C-terminal third of the deduced amino acid sequence.
Host Species: Mouse
Clone: 2A.3
Isotype: IgG1
Cross Reactivity: Human, Cow, Rat, Mouse, and Guinea pig. Others-not known
Epitope: Not determined
Immunogen: Partially purified bovine brain Gαo protein
Molecular Weight: 39-42kDa
Positive Control: IMR-5 cells. Brain
Cellular Localization: Cell membrane
Ab-2 is highly specific to Galphao and shows no cross-reaction with transducin, Gαi1, Gαi2, Gαi3, or Gαs. It reacts equally with the C-terminal splice variants of Gαo (mouse GαoA and GαoB, equivalent to Gαo1 and Gαo2 in other species).
Western Blotting
Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly
Specifications
Specifications
| Antigen | Gαo G-Protein Ab-2 |
| Classification | Monoclonal |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Partially purified bovine brain Gαo protein |
| Quantity | 200 μL |
| Primary or Secondary | Primary |
| Target Species | Bovine, Guinea Pig, Human, Mouse, Rat |
| Form | Purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.