Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAAR3, Polyclonal, Invitrogen™

Catalog No. PIPA132666
Change view
Click to view available options
Quantity:
25 μg
Catalog No. Quantity
PIPA132666 25 μg
1 options

Catalog No. PIPA132666

Supplier: Thermo Scientific PA132666

Rabbit Polyclonal Antibody

PA1-32666 detects GPCR GPR57 from human samples.PA1-32666 has been successfully used in immunohistochemistry (paraffin tissue) applications.The PA1-32666 immunogen is: Synthetic peptide, KLH conjugated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen G Protein-Coupled Rec. 57
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Gene TAAR3P
Gene Accession No. Q9P1P4
Gene Alias GPR57, GPR58, TAAR2
Gene Symbols TAAR3P
Host Species Rabbit
Immunogen Synthetic peptide, KLH conjugated.
Purification Method Antigen affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9288
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.