Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G-protein coupled receptor 56 (GPR56) Mouse anti-Mouse, Clone: H11, MilliporeSigma™

Catalog No. MABN310MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN310MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN310MI Supplier MilliporeSigma Supplier No. MABN310
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-G-protein coupled receptor 56 (GPR56) Antibody, clone H11 is validated for use in Western Blotting, immunocytochemistry, immunohistochemistry for the detection of G-protein coupled receptor 56 (GPR56).
TRUSTED_SUSTAINABILITY

Specifications

Antigen G-protein coupled receptor 56 (GPR56)
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone H11
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_001185823
Gene Symbols GPR56, TM7XN1, TM7LN4, EGF-TM7-like, BFPP
Host Species Mouse
Immunogen N-terminal fragment of mouse GPR56
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.