Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G Protein Giα 1/2 Rabbit anti-Mouse, Rat, Human, Bovine, Polyclonal, MilliporeSigma™

Catalog No. 071500MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-150-0MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-150-0MI Supplier MilliporeSigma Supplier No. 071500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-G Protein Giα 1/2 Antibody is an antibody against G Protein Giα 1/2 for use in western blot, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen G Protein Giα 1/2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal IgG in phosphate buffered saline with 0.08% sodium azide.
Gene Symbols GNAI1; Gi
Host Species Rabbit
Immunogen Peptide corresponding to amino acids 345-354 of Gi1 and 346-355 of Gi2
Purification Method Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_002069.5
Target Species Bovine, Human, Mouse, Rat
Content And Storage -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.