Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G Protein Goα Mouse anti-Bovine, Guinea Pig, Human, Mouse, Rat, Clone: 2A, MilliporeSigma™

Catalog No. MAB3073MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3073MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3073MI Supplier MilliporeSigma Supplier No. MAB3073
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-G Protein Goα Antibody, clone 2A is an antibody against G Protein Goα for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen G Protein Goα
Applications Western Blot
Classification Monoclonal
Clone 2A
Conjugate Unconjugated
Formulation Purified Immunoglobulin. Liquid in 0.02M PBS, 0.25M NaCl, pH 7.6, containing 0.1% sodium azide as a preservative.
Gene Accession No. NM_020988.1; NM_138736.1
Gene Symbols GNAO1, DKFZp686O0962, G-ALPHA-o, GNAO
Host Species Mouse
Immunogen Bovine brain Go-alpha purified by the method of Sternweis and Robishaw (J. Biol. Chem. [1984] 259, 13806).
Quantity 100 μg
Primary or Secondary Primary
Target Species Bovine, Guinea Pig, Human, Mouse, Rat
Content And Storage 2-8°C in undiluted aliquots for up to 6 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.