Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gαq Mouse, Unlabeled, Clone: 10, BD

Catalog No. BDB612704 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612704 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB612704 Supplier BD Biosciences Supplier No. 612704
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The GTP binding regulatory proteins (G proteins) consist of three subunits: α, β, and γ. These heterotrimeric proteins function at membranes to relay signals from cell surface receptors to intracellular effectors. The α subunit is unique for each G protein and contains the site of GTP binding and hydrolysis, as well sites for receptor and effector interactions. The βγ subunit complex interacts directly with receptors and the α subunit. The Gα family includes four families: the Gαs family including Gαs , Gαo1f, and Gαt, the Gαi family including Gαi, Gαo, and Gαz, the Gαq/Gα11 family and the Gα12/13 family. The Gαq protein is 88% homologus with Gα11 and both are widely expressed. These G proteins activate phospholipase C proteins, which induce calcium signaling events. G protein coupled receptors (GPCRs) involved in regulating Wnt signaling activate Gαq, phospholipase Cβ, and induce calcium-dependent activation of calpain. These events promote β-catenin nuclear export and proteolysis. Gαq has also been implicated in metabotropic glutamate receptor signaling. Thus, Gαq isoforms activate phospholipase C proteins in various G-protein coupled receptor pathways.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Gαq
Applications Western Blot
Classification Monoclonal
Clone 10
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Gαq aa. 22-31
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.