Learn More
X-ray repair complementing defective repair in Chinese hamster cells 6, Mouse, Clone: 4C2-1A6, Abnova™
Shop All Abnova Corporation ProductsDescription
The p70/p80 autoantigen is a nuclear complex consisting of two subunits with molecular masses of approximately 70 and 80kDa. The complex functions as a single-stranded DNA-dependent ATP-dependent helicase. The complex may be involved in the repair of nonhomologous DNA ends such as that required for double-strand break repair, transposition, and V(D)J recombination. High levels of autoantibodies to p70 and p80 have been found in some patients with systemic lupus erythematosus. [provided by RefSeq
Specifications
Specifications
| Antigen | X-ray repair complementing defective repair in Chinese hamster cells 6 |
| Applications | ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot |
| Classification | Monoclonal |
| Clone | 4C2-1A6 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against a full length recombinant XRCC6. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | XRCC6 |
| Gene Accession No. | BC008343 |
| Gene Alias | CTC75/CTCBF/G22P1/KU70/ML8/TLAA |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.