Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
G3BP1 Mouse anti-Human, Clone: 14E5-G9, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 051938
Description
Anti-G3BP1 Antibody, clone 14E5-G9 is an antibody against G3BP1 for use in western blotting and immunocytochemistry.
Specifications
G3BP1 | |
Monoclonal | |
Unconjugated | |
NP_005745.1 | |
Mouse | |
Protein G purfied | |
Primary | |
2-8°C for one year | |
IgG1 κ |
Immunocytochemistry, Western Blot | |
14E5-G9 | |
Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide. | |
G3BP, G3BP-1, HDH-VIII | |
GST-tagged recombinant protein corresponding to human G3BP1. | |
100 μg | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction