Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GABA A Receptor α1 chain Mouse anti-Avian, Bovine, Human, Pig, Clone: BD24, MilliporeSigma™

Catalog No. MAB339MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB339MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB339MI Supplier MilliporeSigma Supplier No. MAB339
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-GABA A Receptor α1 chain Antibody, N-terminus, clone BD24 is an antibody against GABA A Receptor α1 chain for use in immunohistochemistry, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GABA A Receptor α1 chain
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone BD24
Conjugate Unconjugated
Formulation Liquid. Buffer = 0.02M Phosphate buffer, 0.25M NaCl with 0.1% sodium azide.
Gene Accession No. NM_000806.4
Gene Symbols GABRA1, ECA4, GABA, EJM
Host Species Mouse
Immunogen Purified GABA/benzodiazepine receptor from bovine cortex.
Quantity 100 μg
Primary or Secondary Primary
Target Species Avian, Bovine, Human, Pig
Content And Storage −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.