Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GABA A Receptor Β2/3 Mouse anti-Bovine, Human, Rat, Clone: 62-3G1, MilliporeSigma™

Catalog No. 05474MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-474-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-474-MI Supplier MilliporeSigma Supplier No. 05474
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-GABA A Receptor β2/3 Antibody, is an antibody against GABA A Receptor β2/3 for use in western blot, immunoprecipitation, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GABA A Receptor β2/3
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 62-3G1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG in buffer containing 0.1MTris-glycine, pH 7.4, 0.15MNaCl, 0.05% sodium azide.
Gene Accession No. XM_612515, XM_583106
Gene Symbols GABRA1, GABA, EJM
Host Species Mouse
Immunogen Affinity-purified GABAA receptor isolated from bovine brain. Clone 62-3G1
Purification Method Protein G purfied
Quantity 200 μg
Primary or Secondary Primary
Target Species Bovine, Human, Rat
Content And Storage 2-8°C for up to 12 months.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.