Learn More
GABA A Receptor Β2 Rabbit anti-Bovine, Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | GABA A Receptor β2 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified immunoglobulin. Liquid in 10mM HEPES, pH 7.5, with 150mM NaCl, 100μg/mL BSA and 50% glycerol. Approximate concentration is 0.2mg/mL. |
| Gene Symbols | GABRB2; MGC119386; GABA; MGC119389; MGC119388 |
| Host Species | Rabbit |
| Immunogen | MBP fusion protein from the cytosolic loop of rat GABAA receptor beta2 subunit. |
| Purification Method | Affinity Purified |
| Quantity | 100 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.