Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GABA A Receptor α4 Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB5459MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5459MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB5459MI Supplier MilliporeSigma Supplier No. AB5459
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-GABA A Receptor α4 Antibody, C-terminus is an antibody against GABA A Receptor α4 for use in immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GABA A Receptor α4
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified immunoglobulin in 10mM HEPES, pH 7.5, with 150mM NaCl, 100μg/mL BSA and 50% glycerol.
Gene Symbols GABRA4; GABA
Host Species Rabbit
Immunogen MBP fusion protein from the cytosolic loop of rat GABA A receptor alpha4 subunit.
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_000809.2
Target Species Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.