Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GABA(A) receptor subunit gamma-2 Mouse anti-Rat, Clone: 10F10-C1-B8, MilliporeSigma™

Catalog No. MABN263MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN263MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN263MI Supplier MilliporeSigma Supplier No. MABN263
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect GABA(A) receptor subunit gamma-2 using this Anti-GABA(A) receptor subunit gamma-2 Antibody, clone 10F10-C1-B8 validated for use in Western Blotting, immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GABA(A) receptor subunit gamma-2
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 10F10-C1-B8
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2bκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_899156
Gene Symbols Gabrg2
Host Species Mouse
Immunogen Linear peptide corresponding to rat GABA(A) receptor subunit gamma-2.
Purification Method Protein G purified
Quantity 100 μL
Primary or Secondary Primary
Target Species Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.