Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GABA Guinea Pig anti-Rat, All, Polyclonal, MilliporeSigma™
Guinea Pig Polyclonal Antibody
Supplier: MilliporeSigma AB175
Description
This Anti-GABA Antibody is validated for use in Immunohistochemistry (Paraffin) and Enzyme Immunoassay (ELISA) for the detection of GABA.
Specifications
GABA | |
Polyclonal | |
Purified guinea pig polyclonal with 0.05% sodium azide. | |
BSA-conjugated GABA-Glutaraldehyde | |
50 μL | |
All species, Rat | |
Serum |
ELISA, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Guinea Pig | |
Depleted serum | |
Primary | |
-20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction