Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GABA B Receptor R2 Guinea Pig anti-Rat, Human, Bovine, Horse, Rhesus Macaque, Chimpanzee, Mouse, Canine, Polyclonal, MilliporeSigma™

Catalog No. AB2255MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB2255MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB2255MI Supplier MilliporeSigma Supplier No. AB2255
Add to Cart
Add to Cart

Guinea Pig Polyclonal Antibody

Anti-GABA B Receptor R2 Antibody is an antibody against GABA B Receptor R2 for use in immunohistochemistry and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GABA B Receptor R2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Unpurified guinea pig polyclonal serum with 0.05% sodium azide.
Gene Symbols GABBR2; GABA-B-R2; GABABR2; GPR51; GPRC3B; Gb2; HG20
Host Species Guinea Pig
Immunogen Bovine thyroglobulin-conjugated linear peptide at the C-terminus of rat GABA-B-R2.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_113990
Target Species Bovine, Canine, Chimpanzee, Equine, Human, Mouse, Rat, Rhesus Monkey
Content And Storage -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.